accessibility menu, dialog, popup

UserName

HHClClH3CNNNNCH3

CAS RN 70775-75-6

IUPAC Name: dihydrogen N-(1-{10-[4-(octylimino)-1,4-dihydropyridin-1-yl]decyl}-1,4-dihydropyridin-4-ylidene)octan-1-amine dichloride
Synonyms: octenidine hcl
Molecular Weight (g/mol): 623.84
Molecular Formula: C36H64Cl2N4
InChi Key: SMGTYJPMKXNQFY-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CCCCCCCCN=C1C=CN(CCCCCCCCCCN2C=CC(C=C2)=NCCCCCCCC)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)