Learn More
Octenidine dihydrochloride, 98%
CAS: 70775-75-6 | C36H64Cl2N4
$71.14 - $254.21
Chemical Identifiers
| CAS | 70775-75-6 |
|---|---|
| Molecular Formula | C36H64Cl2N4 |
| MDL Number | MFCD01938808 |
| Synonym | N,N'-[Decane-1,10-diyldi-1(4H)-pyridyl-4-ylidene]bis(octylammonium) dichloride |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6225703
|
Thermo Scientific Chemicals
H6225703 |
1 g |
Each for $71.14
|
|
|||||
|
AAH6225706
|
Thermo Scientific Chemicals
H6225706 |
5 g |
Each for $254.21
|
|
|||||
Description
Octenidine dihydrochloride is used as a antiseptic for skin, mucous Membranes and wounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsOctenidine dihydrochloride is used as a antiseptic for skin, mucous Membranes and wounds.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
| 70775-75-6 | |
| C36H64Cl2N4 | |
| MFCD01938808 | |
| N,N'-[Decane-1,10-diyldi-1(4H)-pyridyl-4-ylidene]bis(octylammonium) dichloride |
Specifications
| 70775-75-6 | |
| C36H64Cl2N4 | |
| 1 g | |
| N,N'-[Decane-1,10-diyldi-1(4H)-pyridyl-4-ylidene]bis(octylammonium) dichloride | |
| 623.83 | |
| Octenidine dihydrochloride |
| 214°C | |
| MFCD01938808 | |
| 14,6754 | |
| Soluble in dimethyl sulfoxide and methanol. | |
| 98% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 274-861-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only