missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octenidine dihydrochloride, 98%
Description
Octenidine dihydrochloride is used as a antiseptic for skin, mucous Membranes and wounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Octenidine dihydrochloride is used as a antiseptic for skin, mucous Membranes and wounds.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Octenidine dihydrochloride is used as a antiseptic for skin, mucous Membranes and wounds.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 214°C |
| Quantity | 1 g |
| Merck Index | 14,6754 |
| Solubility Information | Soluble in dimethyl sulfoxide and methanol. |
| Formula Weight | 623.83 |
| Percent Purity | 98% |
| Chemical Name or Material | Octenidine dihydrochloride |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 274-861-8 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only