missing translation for 'onlineSavingsMsg'
Learn More

Octaphenylcyclotetrasiloxane, 98+%

Catalog No. aal0613022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0613022 100 g
AAL0613014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0613022 Supplier Thermo Scientific Chemicals Supplier No. L0613022
Only null left

CAS: 546-56-5 | C48H40O4Si4 | 793.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 546-56-5
Molecular Formula C48H40O4Si4
Molecular Weight (g/mol) 793.18
MDL Number MFCD00003268
InChI Key VSIKJPJINIDELZ-UHFFFAOYSA-N
Synonym octaphenylcyclotetrasiloxane, cyclotetrasiloxane, octaphenyl, 2,2,4,4,6,6,8,8-octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, octaphenyltetracyclosiloxane, cyclotetrasiloxane, 2,2,4,4,6,6,8,8-octaphenyl, 1,1,3,3,5,5,7,7-octaphenylcyclotetrasiloxane, octaphenyl tetracyclosiloxane, acmc-1aqwg, ph2sio 4, vsikjpjinidelz-uhfffaoysa
PubChem CID 68347
IUPAC Name 2,2,4,4,6,6,8,8-octakis-phenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
SMILES O1[Si](O[Si](O[Si](O[Si]1(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 332°C (1mmHg)
Melting Point 193°C to 200°C
Quantity 100 g
Beilstein 2320758
Formula Weight 793.19
Percent Purity ≥98%
Chemical Name or Material Octaphenylcyclotetrasiloxane
EINECSNumber 208-904-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only