missing translation for 'onlineSavingsMsg'
Learn More

Octanoyl chloride, 99%

Catalog No. p-177003
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC129431000 100 g Glass bottle
AC129435000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC129431000 Supplier Thermo Scientific Chemicals Supplier No. 129431000
Only null left

CAS: 111-64-8 | C8H15ClO | 162.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-64-8
MDL Number MFCD00000766
InChI Key REEZZSHJLXOIHL-UHFFFAOYSA-N
Synonym capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride
PubChem CID 8124
IUPAC Name octanoyl chloride
SMILES CCCCCCCC(=O)Cl

Specifications

Acidity 1% max.
CAS Min % 98.5
CAS Max % 100.0
Melting Point -63.0°C
Density 0.9500g/mL
Boiling Point 195.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H15ClO
Refractive Index 1.4340 to 1.4360
Linear Formula CH3(CH2)6COCl
Quantity 100 g
Beilstein 02, 348
Specific Gravity 0.95
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohol, miscible with most common organic solvents, soluble in ether
Molecular Weight (g/mol) 162.66
Viscosity 1.51 mPa.s (20°C)
Formula Weight 162.66
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Octanoyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye damage.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a dry place.
Store in a closed container.
IF IN EYES: Ri
EINECSNumber 203-891-6
RTECSNumber RH1570000
TSCA TSCA

RUO – Research Use Only