missing translation for 'onlineSavingsMsg'
Learn More

Octanoyl chloride, 99%

Catalog No. AAL03526AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL03526AE 100 mL
AAL03526AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL03526AE Supplier Thermo Scientific Chemicals Supplier No. L03526AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 111-64-8 | C8H15ClO | 162.657 g/mol

Octanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.

Solubility
Soluble in organic solvents (acetone, chloroform, toluene, THF). Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with Bases, Alcohols, Amines, Oxidizing agents. Store away from alcohol and amines.

Chemical Identifiers

CAS 111-64-8
Molecular Formula C8H15ClO
Molecular Weight (g/mol) 162.657
MDL Number MFCD00000766
InChI Key REEZZSHJLXOIHL-UHFFFAOYSA-N
Synonym capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride
PubChem CID 8124
IUPAC Name octanoyl chloride
SMILES CCCCCCCC(=O)Cl

Specifications

Melting Point -63°C
Density 0.942
Boiling Point 195°C
Flash Point 80°C (176°F)
Odor Pungent
Refractive Index 1.435
Quantity 100 mL
UN Number UN3265
Beilstein 635917
Sensitivity Moisture sensitive
Solubility Information Soluble in organic solvents (acetone,chloroform,toluene,THF). Hydrolyzes in water.
Formula Weight 162.66
Percent Purity 99%
Chemical Name or Material Octanoyl chloride
Show More Show Less
Hazard Category H227-H290-H315-H317-H318-H330
Hazard Statement GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P233-P234-P240-P241-P242-P243-P260-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P310-P333+P313-P362-P374-P390-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 203-891-6
RTECSNumber RH1570000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only