accessibility menu, dialog, popup

UserName

H3CO

CAS RN 1674-37-9

IUPAC Name: 1-phenyloctan-1-one
Synonyms: 1-octanone, 1-phenyl-
Molecular Weight (g/mol): 204.31
Molecular Formula: C14H20O
InChi Key: UDEVCZRUNOLVLU-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)