missing translation for 'onlineSavingsMsg'
Learn More

Octanophenone, 99+%

Catalog No. AC279360050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC279360050 5 g Glass Bottle
AC279360250 25 g Glass Bottle
AC279361000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC279360050 Supplier Thermo Scientific Chemicals Supplier No. 279360050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1674-37-9 | C14H20O | 204.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1674-37-9
Molecular Formula C14H20O
Molecular Weight (g/mol) 204.31
MDL Number MFCD00003554
InChI Key UDEVCZRUNOLVLU-UHFFFAOYSA-N
Synonym octanophenone, heptyl phenyl ketone, 1-octanone, 1-phenyl, caprylophenone, n-octanophenone, ketone, heptyl phenyl, 4-n-butylbutyrophenone, 1-phenyl-octan-1-one, acmc-1axlq
PubChem CID 74291
IUPAC Name 1-phenyloctan-1-one
SMILES CCCCCCCC(=O)C1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.93g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Refractive Index 1.5024 to 1.5054 (20°C, 589nm)
Linear Formula C6H5CO(CH2)6CH3
Quantity 5 g
Formula Weight 204.31
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Octanophenone
Show More Show Less

RUO – Research Use Only