missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octanophenone, 98%
Description
Octanophenone is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Octanophenone is used as a pharmaceutical intermediate.
Solubility
Soluble in hot ethanol, acetone, toluene and other common organic solvents. Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Octanophenone is used as a pharmaceutical intermediate.
Solubility
Soluble in hot ethanol, acetone, toluene and other common organic solvents. Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 22°C to 24°C |
| Density | 0.938 |
| Boiling Point | 285°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.504 |
| Quantity | 5 g |
| Beilstein | 1868534 |
| Solubility Information | Soluble in hot ethanol,acetone,toluene and other common organic solvents. Insoluble in water. |
| Formula Weight | 204.31 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| EINECSNumber | 216-817-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only