missing translation for 'onlineSavingsMsg'
Learn More

Octanophenone, 98%

Catalog No. AAA1365206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1365206 5 g
AAA1365214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1365206 Supplier Thermo Scientific Chemicals Supplier No. A1365206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1674-37-9 | C14H20O | 204.31 g/mol

Octanophenone is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanophenone is used as a pharmaceutical intermediate.

Solubility
Soluble in hot ethanol, acetone, toluene and other common organic solvents. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1674-37-9
Molecular Formula C14H20O
Molecular Weight (g/mol) 204.31
MDL Number MFCD00003554
InChI Key UDEVCZRUNOLVLU-UHFFFAOYSA-N
Synonym octanophenone, heptyl phenyl ketone, 1-octanone, 1-phenyl, caprylophenone, n-octanophenone, ketone, heptyl phenyl, 4-n-butylbutyrophenone, 1-phenyl-octan-1-one, acmc-1axlq
PubChem CID 74291
IUPAC Name 1-phenyloctan-1-one
SMILES CCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 22°C to 24°C
Density 0.938
Boiling Point 285°C
Flash Point >110°C (230°F)
Refractive Index 1.504
Quantity 5 g
Beilstein 1868534
Solubility Information Soluble in hot ethanol,acetone,toluene and other common organic solvents. Insoluble in water.
Formula Weight 204.31
Percent Purity 98%
Chemical Name or Material Octanophenone
Show More Show Less
EINECSNumber 216-817-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only