missing translation for 'onlineSavingsMsg'
Learn More

Octanohydroxamic Acid 99.0+%, TCI America™

Catalog No. C12325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12325G 5 g
C123225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12325G Supplier TCI America Supplier No. C12325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7377-03-9
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00143918
InChI Key RGUVUPQQFXCJFC-UHFFFAOYSA-N
Synonym Caprylohydroxamic Acid, N-Hydroxyoctanamide
PubChem CID 23846
IUPAC Name N-hydroxyoctanamide
SMILES CCCCCCCC(=O)NO

Specifications

Melting Point 78°C
Color White
Quantity 5 g
Formula Weight 159.23
Percent Purity ≥99.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Octanohydroxamic Acid
RTECSNumber RG9900500
TSCA Yes