accessibility menu, dialog, popup

UserName

H3CONHOH

CAS RN 7377-03-9

IUPAC Name: N-hydroxyoctanamide
Synonyms: caprylhydroxamic acid octanohydroxamic acid
Molecular Weight (g/mol): 159.23
Molecular Formula: C8H17NO2
InChi Key: RGUVUPQQFXCJFC-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)NO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results