missing translation for 'onlineSavingsMsg'
Learn More

Octanal, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019874
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019874 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019874 Supplier Merck Emd Millipore Supplier No. 524661ML
Only null left
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 124-13-0
Vapor Pressure 2 mmHg (20°C)
Density 0.82 g/mL (at 25°C)
Flash Point 52°C
Physical Form Neat
Linear Formula CH3(CH2)6 CHO
MDL Number MFCD00007029
Melting Point 12°C to 15°C
Boiling Point 171°C
Synonym Aldehyde C8; Caprylic aldehyde; Octyl aldehyde
Type Analytical Standard
Quantity 1 mL
Formula Weight 128.21
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less