missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octamethylcyclotetrasiloxane 98.0+%, TCI America™
Supplier: TCI America O0142100ML
Specifications
| Octamethylcyclotetrasiloxane | |
| Colorless | |
| C8H24O4Si4 | |
| 100 mL | |
| octamethylcyclotetrasiloxane, cyclotetrasiloxane, octamethyl, oktamethylcyklotetrasiloxan, cyclic dimethylsiloxane tetramer, omcts, nuc silicone vs 7207, union carbide 7207, silicone sf 1173, oktamethylzyklotetrasiloxan, unii-cz227117je | |
| C[Si]1(O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)C | |
| 296.616 | |
| CHEBI:25640 | |
| ≥98.0% (GC) |
| 556-67-2 | |
| 175°C | |
| MFCD00003269 | |
| 1992 | |
| HMMGMWAXVFQUOA-UHFFFAOYSA-N | |
| 2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane | |
| 11169 | |
| 296.62 | |
| Liquid |
Chemical Identifiers
| 556-67-2 | |
| 296.616 | |
| HMMGMWAXVFQUOA-UHFFFAOYSA-N | |
| 11169 | |
| 2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane |
| C8H24O4Si4 | |
| MFCD00003269 | |
| octamethylcyclotetrasiloxane, cyclotetrasiloxane, octamethyl, oktamethylcyklotetrasiloxan, cyclic dimethylsiloxane tetramer, omcts, nuc silicone vs 7207, union carbide 7207, silicone sf 1173, oktamethylzyklotetrasiloxan, unii-cz227117je | |
| CHEBI:25640 | |
| C[Si]1(O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)C |
Safety and Handling
EINECSNumber : (7)-0475
RTECSNumber : GZ4397000
TSCA : Yes