missing translation for 'onlineSavingsMsg'
Learn More

Octamethylcyclotetrasiloxane 98.0+%, TCI America™

Catalog No. p-7134205
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O014225ML 25 mL
O0142100ML 100 mL
O0142500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O014225ML Supplier TCI America Supplier No. O014225ML
Only null left

Chemical Identifiers

CAS 556-67-2
Molecular Formula C8H24O4Si4
Molecular Weight (g/mol) 296.616
MDL Number MFCD00003269
InChI Key HMMGMWAXVFQUOA-UHFFFAOYSA-N
Synonym octamethylcyclotetrasiloxane, cyclotetrasiloxane, octamethyl, oktamethylcyklotetrasiloxan, cyclic dimethylsiloxane tetramer, omcts, nuc silicone vs 7207, union carbide 7207, silicone sf 1173, oktamethylzyklotetrasiloxan, unii-cz227117je
PubChem CID 11169
ChEBI CHEBI:25640
IUPAC Name 2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
SMILES C[Si]1(O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)C

Specifications

Color Colorless
Boiling Point 175°C
Quantity 25 mL
UN Number 1992
Formula Weight 296.62
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Octamethylcyclotetrasiloxane
EINECSNumber (7)-0475
RTECSNumber GZ4397000
TSCA Yes