missing translation for 'onlineSavingsMsg'
Learn More

Octafluoronaphthalene, 96%

Catalog No. AAB2169903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2169903 1 g
AAB2169906 5 g
AAB2169914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2169903 Supplier Thermo Scientific Chemicals Supplier No. B2169903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 313-72-4 | C10F8 | 272.097 g/mol

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water. Solubility in toluene, almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 313-72-4
Molecular Formula C10F8
Molecular Weight (g/mol) 272.097
MDL Number MFCD00014307
InChI Key JDCMOHAFGDQQJX-UHFFFAOYSA-N
Synonym octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion
PubChem CID 67564
IUPAC Name 1,2,3,4,5,6,7,8-octafluoronaphthalene
SMILES C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F

Specifications

Melting Point 84°C to 89°C
Boiling Point 209°C
Quantity 1 g
Beilstein 1915630
Solubility Information Insoluble in water. Solubility in toluene,almost transparent.
Formula Weight 272.1
Percent Purity 96%
Chemical Name or Material Octafluoronaphthalene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-239-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only