missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octafluoroadipic Acid 98.0+%, TCI America™
$167.08 - $503.13
Chemical Identifiers
| CAS | 336-08-3 |
|---|---|
| Molecular Formula | C6H2F8O4 |
| Molecular Weight (g/mol) | 290.07 |
| MDL Number | MFCD00066405 |
| InChI Key | AXRSOGFYDSXLQX-UHFFFAOYSA-N |
| Synonym | Octafluorohexanedioic Acid, Perfluoroadipic Acid, Perfluorohexanedioic Acid |
| PubChem CID | 67640 |
| IUPAC Name | octafluorohexanedioic acid |
| SMILES | OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O |
Chemical Identifiers
| 336-08-3 | |
| 290.07 | |
| AXRSOGFYDSXLQX-UHFFFAOYSA-N | |
| 67640 | |
| OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O |
| C6H2F8O4 | |
| MFCD00066405 | |
| Octafluorohexanedioic Acid, Perfluoroadipic Acid, Perfluorohexanedioic Acid | |
| octafluorohexanedioic acid |
Specifications
| 336-08-3 | |
| White-Yellow | |
| MFCD00066405 | |
| Octafluorohexanedioic Acid, Perfluoroadipic Acid, Perfluorohexanedioic Acid | |
| OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O | |
| 290.07 | |
| 290.07 | |
| Crystalline Powder |
| 134°C | |
| C6H2F8O4 | |
| 5 g | |
| AXRSOGFYDSXLQX-UHFFFAOYSA-N | |
| octafluorohexanedioic acid | |
| 67640 | |
| ≥98.0% (GC,T) | |
| Octafluoroadipic Acid |
Safety and Handling
TSCA : Yes