missing translation for 'onlineSavingsMsg'
Learn More

Octafluoroadipic Acid 98.0+%, TCI America™

Supplier:  TCI America O026025G

 View more versions of this product

Catalog No. O026025G


Only null left

Description

Description

Specifications

Specifications

Octafluoroadipic Acid
134°C
C6H2F8O4
25 g
AXRSOGFYDSXLQX-UHFFFAOYSA-N
octafluorohexanedioic acid
67640
≥98.0% (GC,T)
336-08-3
White-Yellow
MFCD00066405
Octafluorohexanedioic Acid, Perfluoroadipic Acid, Perfluorohexanedioic Acid
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O
290.07
290.07
Crystalline Powder

Chemical Identifiers

336-08-3
290.07
AXRSOGFYDSXLQX-UHFFFAOYSA-N
67640
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O
C6H2F8O4
MFCD00066405
Octafluorohexanedioic Acid, Perfluoroadipic Acid, Perfluorohexanedioic Acid
octafluorohexanedioic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions