accessibility menu, dialog, popup

UserName

H3CSiOCH3OCH3OCH3

CAS RN 3069-42-9

IUPAC Name: trimethoxy(octadecyl)silane
Synonyms: octadecyltrimethoxysilane
Molecular Weight (g/mol): 374.68
Molecular Formula: C21H46O3Si
InChi Key: SLYCYWCVSGPDFR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC[Si](OC)(OC)OC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)