missing translation for 'onlineSavingsMsg'
Learn More

Octadecylphosphonic Acid 98.0+%, TCI America™

Catalog No. O03715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03715G 5 g
O03711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03715G Supplier TCI America Supplier No. O03715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4724-47-4
Molecular Formula C18H39O3P
Molecular Weight (g/mol) 334.481
MDL Number MFCD00015839
InChI Key FTMKAMVLFVRZQX-UHFFFAOYSA-N
Synonym n-octadecylphosphonic acid, phosphonic acid, octadecyl, phosphonic acid, p-octadecyl, n-octadecylphosphonicacid, octadecanephosphonic acid, octadecyl phosphonic acid, octadecyl-phosphonic acid, acmc-209k8h, ksc492m8n, octadecylphosphonic acid
PubChem CID 78451
IUPAC Name octadecylphosphonic acid
SMILES CCCCCCCCCCCCCCCCCCP(=O)(O)O

Specifications

Melting Point 99°C
Color White
Quantity 5 g
Formula Weight 334.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Octadecylphosphonic Acid
EINECSNumber (2)-3860
TSCA Yes