missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt 97.0+%, TCI America™
$57.79 - $296.38
Chemical Identifiers
| CAS | 13177-41-8 |
|---|---|
| Molecular Formula | C23H49NO3S |
| Molecular Weight (g/mol) | 419.709 |
| MDL Number | MFCD00043218 |
| InChI Key | DIROHOMJLWMERM-UHFFFAOYSA-N |
| Synonym | 3-[Dimethyl(octadecyl)ammonio]propane-1-sulfonate, 3-[Dimethyl(stearyl)ammonio]propane-1-sulfonate, Stearyl Sulfobetaine, Stearyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt |
| PubChem CID | 83205 |
| IUPAC Name | 3-[dimethyl(octadecyl)azaniumyl]propane-1-sulfonate |
| SMILES | CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
Chemical Identifiers
| 13177-41-8 | |
| 419.709 | |
| DIROHOMJLWMERM-UHFFFAOYSA-N | |
| 83205 | |
| CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
| C23H49NO3S | |
| MFCD00043218 | |
| 3-[Dimethyl(octadecyl)ammonio]propane-1-sulfonate, 3-[Dimethyl(stearyl)ammonio]propane-1-sulfonate, Stearyl Sulfobetaine, Stearyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt | |
| 3-[dimethyl(octadecyl)azaniumyl]propane-1-sulfonate |
Specifications
| 13177-41-8 | |
| White | |
| MFCD00043218 | |
| 3-[Dimethyl(octadecyl)ammonio]propane-1-sulfonate, 3-[Dimethyl(stearyl)ammonio]propane-1-sulfonate, Stearyl Sulfobetaine, Stearyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt | |
| CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] | |
| 419.709 | |
| 419.71 | |
| Crystalline Powder |
| 260°C | |
| C23H49NO3S | |
| 25 g | |
| DIROHOMJLWMERM-UHFFFAOYSA-N | |
| 3-[dimethyl(octadecyl)azaniumyl]propane-1-sulfonate | |
| 83205 | |
| ≥97.0% (N) | |
| Octadecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt |
Safety and Handling
TSCA : Yes