accessibility menu, dialog, popup

UserName

CAS RN 13177-41-8

IUPAC Name: 3-[dimethyl(octadecyl)azaniumyl]propane-1-sulfonate
Synonyms: 3-[dimethyl(octadecyl)ammonio]propane-1-sulfonate
Molecular Weight (g/mol): 419.71
Molecular Formula: C23H49NO3S
InChi Key: DIROHOMJLWMERM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results