missing translation for 'onlineSavingsMsg'
Learn More

Octadecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt 97.0+%, TCI America™

Catalog No. O026125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O026125G 25 g
O0261250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O026125G Supplier TCI America Supplier No. O026125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13177-41-8
Molecular Formula C23H49NO3S
Molecular Weight (g/mol) 419.709
MDL Number MFCD00043218
InChI Key DIROHOMJLWMERM-UHFFFAOYSA-N
Synonym 3-[Dimethyl(octadecyl)ammonio]propane-1-sulfonate, 3-[Dimethyl(stearyl)ammonio]propane-1-sulfonate, Stearyl Sulfobetaine, Stearyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt
PubChem CID 83205
IUPAC Name 3-[dimethyl(octadecyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]

Specifications

Melting Point 260°C
Color White
Quantity 25 g
Formula Weight 419.71
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Octadecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt