missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecyl Vinyl Ether 85.0+%, TCI America™
$68.97 - $452.97
Chemical Identifiers
| CAS | 930-02-9 |
|---|---|
| Molecular Formula | C20H40O |
| Molecular Weight (g/mol) | 296.539 |
| MDL Number | MFCD00026686 |
| InChI Key | QJJDJWUCRAPCOL-UHFFFAOYSA-N |
| Synonym | Stearyl Vinyl Ether |
| PubChem CID | 13585 |
| IUPAC Name | 1-ethenoxyoctadecane |
| SMILES | CCCCCCCCCCCCCCCCCCOC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
O001925ML
|
TCI America
O001925ML |
25 mL |
Each for $68.97
|
|
|||||
|
O0019500ML
|
TCI America
O0019500ML |
500 mL |
Each for $452.97
|
|
|||||
Chemical Identifiers
| 930-02-9 | |
| 296.539 | |
| QJJDJWUCRAPCOL-UHFFFAOYSA-N | |
| 13585 | |
| CCCCCCCCCCCCCCCCCCOC=C |
| C20H40O | |
| MFCD00026686 | |
| Stearyl Vinyl Ether | |
| 1-ethenoxyoctadecane |
Specifications
| 930-02-9 | |
| C20H40O | |
| 25 mL | |
| QJJDJWUCRAPCOL-UHFFFAOYSA-N | |
| 1-ethenoxyoctadecane | |
| 13585 | |
| ≥85.0% (GC) | |
| Octadecyl Vinyl Ether |
| White-Yellow | |
| MFCD00026686 | |
| Stearyl Vinyl Ether | |
| CCCCCCCCCCCCCCCCCCOC=C | |
| 296.539 | |
| 296.54 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-0372&(2)-0575
RTECSNumber : RG0300000
TSCA : Yes