missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecyl Vinyl Ether 85.0+%, TCI America™
Supplier: TCI America O0019500ML
Specifications
| Octadecyl Vinyl Ether | |
| White-Yellow | |
| MFCD00026686 | |
| Stearyl Vinyl Ether | |
| CCCCCCCCCCCCCCCCCCOC=C | |
| 296.539 | |
| 296.54 | |
| Crystalline Lumps |
| 930-02-9 | |
| C20H40O | |
| 500 mL | |
| QJJDJWUCRAPCOL-UHFFFAOYSA-N | |
| 1-ethenoxyoctadecane | |
| 13585 | |
| ≥85.0% (GC) |
Chemical Identifiers
| 930-02-9 | |
| 296.539 | |
| QJJDJWUCRAPCOL-UHFFFAOYSA-N | |
| 13585 | |
| CCCCCCCCCCCCCCCCCCOC=C |
| C20H40O | |
| MFCD00026686 | |
| Stearyl Vinyl Ether | |
| 1-ethenoxyoctadecane |
Safety and Handling
EINECSNumber : (2)-0372&(2)-0575
RTECSNumber : RG0300000
TSCA : Yes