missing translation for 'onlineSavingsMsg'
Learn More

Octadecyl Sulfide 96.0+%, TCI America™

Catalog No. O014825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O014825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O014825G Supplier TCI America Supplier No. O014825G
Only null left

Chemical Identifiers

CAS 1844-09-3
Molecular Formula C36H74S
Molecular Weight (g/mol) 539.048
MDL Number MFCD00026688
InChI Key IHWDIGHWDQPQMQ-UHFFFAOYSA-N
Synonym Dioctadecyl Sulfide, Stearyl Sulfide
PubChem CID 74609
IUPAC Name 1-octadecylsulfanyloctadecane
SMILES CCCCCCCCCCCCCCCCCCSCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 70°C
Color White
Quantity 25 g
Formula Weight 539.05
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Octadecyl Sulfide
EINECSNumber (2)-0473&(2)-0475
TSCA Yes