missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecyl Isocyanate 80.0+%, TCI America™
$83.89 - $257.24
Chemical Identifiers
| CAS | 112-96-9 |
|---|---|
| Molecular Formula | C19H37NO |
| Molecular Weight (g/mol) | 295.51 |
| MDL Number | MFCD00002041 |
| InChI Key | QWDQYHPOSSHSAW-UHFFFAOYSA-N |
| Synonym | Isocyanic Acid Octadecyl Ester, Isocyanic Acid Stearyl Ester, Stearyl Isocyanate |
| PubChem CID | 66988 |
| IUPAC Name | 1-isocyanatooctadecane |
| SMILES | CCCCCCCCCCCCCCCCCCN=C=O |
Chemical Identifiers
| 112-96-9 | |
| 295.51 | |
| QWDQYHPOSSHSAW-UHFFFAOYSA-N | |
| 66988 | |
| CCCCCCCCCCCCCCCCCCN=C=O |
| C19H37NO | |
| MFCD00002041 | |
| Isocyanic Acid Octadecyl Ester, Isocyanic Acid Stearyl Ester, Stearyl Isocyanate | |
| 1-isocyanatooctadecane |
Specifications
| 112-96-9 | |
| 220°C | |
| MFCD00002041 | |
| 2206 | |
| QWDQYHPOSSHSAW-UHFFFAOYSA-N | |
| 1-isocyanatooctadecane | |
| 66988 | |
| ≥80.0% (GC) | |
| Octadecyl Isocyanate |
| Yellow | |
| C19H37NO | |
| 25 g | |
| Isocyanic Acid Octadecyl Ester, Isocyanic Acid Stearyl Ester, Stearyl Isocyanate | |
| CCCCCCCCCCCCCCCCCCN=C=O | |
| 295.51 | |
| 295.51 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1692
RTECSNumber : NR0140000
TSCA : Yes
Recommended Storage : Refrigerator