missing translation for 'onlineSavingsMsg'
Learn More

Octadecyl Isocyanate 80.0+%, TCI America™

Catalog No. I013025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I013025G 25 g
I0130250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I013025G Supplier TCI America Supplier No. I013025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-96-9
Molecular Formula C19H37NO
Molecular Weight (g/mol) 295.51
MDL Number MFCD00002041
InChI Key QWDQYHPOSSHSAW-UHFFFAOYSA-N
Synonym Isocyanic Acid Octadecyl Ester, Isocyanic Acid Stearyl Ester, Stearyl Isocyanate
PubChem CID 66988
IUPAC Name 1-isocyanatooctadecane
SMILES CCCCCCCCCCCCCCCCCCN=C=O

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 g
UN Number 2206
Formula Weight 295.51
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Octadecyl Isocyanate
EINECSNumber (2)-1692
RTECSNumber NR0140000
TSCA Yes
Recommended Storage Refrigerator