missing translation for 'onlineSavingsMsg'
Learn More

Octadecyl 3-Mercaptopropionate (contains ca. 12% Hexadecyl 3-Mercaptopropionate) 85.0+%, TCI America™

Catalog No. p-7134295
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O036225G 25 g
O0362500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O036225G Supplier TCI America Supplier No. O036225G

Chemical Identifiers

CAS 31778-15-1
Molecular Formula C21H42O2S
Molecular Weight (g/mol) 358.63
MDL Number MFCD00046844
InChI Key UCUPUEARJPTGKU-UHFFFAOYSA-N
Synonym 3-Mercaptopropionic Acid Octadecyl Ester, Stearyl 3-Mercaptopropionate, 3-Mercaptopropionic Acid Stearyl Ester
PubChem CID 169361
IUPAC Name octadecyl 3-sulfanylpropanoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)CCS

Specifications

Color White
Quantity 25 g
Formula Weight 358.63
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Octadecyl 3-Mercaptopropionate (contains ca. 12% Hexadecyl 3-Mercaptopropionate)
EINECSNumber (2)-1383
TSCA Yes