accessibility menu, dialog, popup

UserName

CAS RN 31778-15-1

H3COOSH

CAS RN 31778-15-1

IUPAC Name: octadecyl 3-sulfanylpropanoate
Synonyms: octadecyl 3-sulfanylpropanoate
Molecular Weight (g/mol): 358.63
Molecular Formula: C21H42O2S
InChi Key: UCUPUEARJPTGKU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOC(=O)CCS
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results