missing translation for 'onlineSavingsMsg'
Learn More

Octadecenylsuccinic Anhydride (mixture of isomers) 93.0+%, TCI America™

Catalog No. O001325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O001325G 25 g
O0013500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O001325G Supplier TCI America Supplier No. O001325G

Chemical Identifiers

CAS 28777-98-2
Molecular Formula C22H38O3
Molecular Weight (g/mol) 350.543
MDL Number MFCD00014545
InChI Key KAYAKFYASWYOEB-ISLYRVAYSA-N
Synonym 3-octadec-1-en-1-yl dihydrofuran-2,5-dione, 3-1e-octadec-1-en-1-yl oxolane-2,5-dione, n-octadecenylsuccinic anhydride, dsstox_cid_7948, dsstox_rid_78620, dsstox_gsid_27948, n-octadecenyl succinic anhydride, 3-e-octadec-1-enyl oxolane-2,5-dione, 2,5-furandione, dihydro-3-1-octadecen-1-yl, n-octadecenylsuccinic anhydride, mixture of isomers
PubChem CID 5354626
IUPAC Name 3-[(E)-octadec-1-enyl]oxolane-2,5-dione
SMILES CCCCCCCCCCCCCCCCC=CC1CC(=O)OC1=O

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 350.54
Percent Purity ≥93.0% (GC)
Chemical Name or Material Octadecenylsuccinic Anhydride (mixture of isomers)
EINECSNumber (2)-0852
RTECSNumber LU1245000
TSCA Yes