accessibility menu, dialog, popup

UserName

H2COOO

CAS RN 28777-98-2

IUPAC Name: 3-(octadec-17-en-1-yl)oxolane-2,5-dione
Synonyms: 3-(octadec-17-en-1-yl)oxolane-2,5-dione
Molecular Weight (g/mol): 350.54
Molecular Formula: C22H38O3
InChi Key: UPIDLHWAQXLUSM-UHFFFAOYNA-N
SMILES: C=CCCCCCCCCCCCCCCCCC1CC(=O)OC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results