missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecanophenone 98.0+%, TCI America™
Supplier: TCI America O018525G
Specifications
| Octadecanophenone | |
| 64°C | |
| 243°C | |
| MFCD00010700 | |
| octadecanophenone, stearophenone, n-octadecanophenone, heptadecyl phenyl ketone, 1-octadecanone, 1-phenyl, 1-phenyl-1-octadecanone # | |
| CCCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1 | |
| 344.583 | |
| 344.58 | |
| Crystalline Powder |
| 6786-36-3 | |
| White | |
| C24H40O | |
| 25 g | |
| DBLXXVQTWJFJFI-UHFFFAOYSA-N | |
| 1-phenyloctadecan-1-one | |
| 81244 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 6786-36-3 | |
| 344.583 | |
| DBLXXVQTWJFJFI-UHFFFAOYSA-N | |
| 81244 | |
| CCCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1 |
| C24H40O | |
| MFCD00010700 | |
| octadecanophenone, stearophenone, n-octadecanophenone, heptadecyl phenyl ketone, 1-octadecanone, 1-phenyl, 1-phenyl-1-octadecanone # | |
| 1-phenyloctadecan-1-one |
Safety and Handling
TSCA : No