missing translation for 'onlineSavingsMsg'
Learn More

Octadecanophenone 98.0+%, TCI America™

Catalog No. O01855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01855G 5 g
O018525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01855G Supplier TCI America Supplier No. O01855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6786-36-3
Molecular Formula C24H40O
Molecular Weight (g/mol) 344.583
MDL Number MFCD00010700
InChI Key DBLXXVQTWJFJFI-UHFFFAOYSA-N
Synonym octadecanophenone, stearophenone, n-octadecanophenone, heptadecyl phenyl ketone, 1-octadecanone, 1-phenyl, 1-phenyl-1-octadecanone #
PubChem CID 81244
IUPAC Name 1-phenyloctadecan-1-one
SMILES CCCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 64°C
Color White
Boiling Point 243°C
Quantity 5 g
Formula Weight 344.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Octadecanophenone