missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecane 98.0+%, TCI America™
$37.29 - $206.90
Chemical Identifiers
| CAS | 593-45-3 |
|---|---|
| Molecular Formula | C18H38 |
| Molecular Weight (g/mol) | 254.502 |
| MDL Number | MFCD00009007 |
| InChI Key | RZJRJXONCZWCBN-UHFFFAOYSA-N |
| Synonym | n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group |
| PubChem CID | 11635 |
| ChEBI | CHEBI:32926 |
| IUPAC Name | octadecane |
| SMILES | CCCCCCCCCCCCCCCCCC |
Chemical Identifiers
| 593-45-3 | |
| 254.502 | |
| RZJRJXONCZWCBN-UHFFFAOYSA-N | |
| 11635 | |
| octadecane |
| C18H38 | |
| MFCD00009007 | |
| n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group | |
| CHEBI:32926 | |
| CCCCCCCCCCCCCCCCCC |
Specifications
| 593-45-3 | |
| C18H38 | |
| 25 g | |
| RZJRJXONCZWCBN-UHFFFAOYSA-N | |
| octadecane | |
| 11635 | |
| 254.50 | |
| Crystalline Lumps |
| White | |
| MFCD00009007 | |
| n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group | |
| CCCCCCCCCCCCCCCCCC | |
| 254.502 | |
| CHEBI:32926 | |
| ≥98.0% (GC) | |
| Octadecane |
Safety and Handling
EINECSNumber : (2)-0010
TSCA : Yes