accessibility menu, dialog, popup

UserName

CAS RN 593-45-3

IUPAC Name: octadecane
Synonyms: octadecane 1-(14)C-labeled cpd octadecane 1-(14)c-labeled cpd octadecane N-octadecane 14C-labeled cpd octadecane ccris 681 octadecane, 14C-labeled cpd octadecane, 14c-labeled cpd 14c-labeled cpd octadecane oktadekan
Molecular Weight (g/mol): 254.5
Molecular Formula: C18H38
InChi Key: RZJRJXONCZWCBN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (8)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (6)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (8)
  • (3)
  • (8)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)
  • (2)