Learn More
Octadecane, 99+%
CAS: 593-45-3 | C18H38 | 254.5 g/mol
$52.95 - $806.43
Chemical Identifiers
| CAS | 593-45-3 |
|---|---|
| Molecular Formula | C18H38 |
| Molecular Weight (g/mol) | 254.5 |
| InChI Key | RZJRJXONCZWCBN-UHFFFAOYSA-N |
| Synonym | n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group |
| PubChem CID | 11635 |
| ChEBI | CHEBI:32926 |
| IUPAC Name | octadecane |
| SMILES | CCCCCCCCCCCCCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC129290050
|
Thermo Scientific Chemicals
129290050 |
5 mL | Glass bottle |
Each for $52.95
|
|
||||
|
AC129292500
|
Thermo Scientific Chemicals
129292500 |
250 mL | Glass bottle |
Each for $235.67
|
|
||||
|
AC129290010
|
Thermo Scientific Chemicals
129290010 |
1 L | Glass bottle |
Each for $806.43
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 593-45-3 | |
| 254.5 | |
| n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group | |
| CHEBI:32926 | |
| CCCCCCCCCCCCCCCCCC |
| C18H38 | |
| RZJRJXONCZWCBN-UHFFFAOYSA-N | |
| 11635 | |
| octadecane |
Specifications
| 593-45-3 | |
| White | |
| 317.0°C | |
| Authentic | |
| Glass bottle | |
| CH3(CH2)16CH3 | |
| 01,173 | |
| n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group | |
| RZJRJXONCZWCBN-UHFFFAOYSA-N | |
| octadecane | |
| 11635 | |
| 254.5 | |
| Crystalline Low Melting Solid |
| 28.0°C to 31.0°C | |
| 0.7770g/mL | |
| 166°C | |
| 99% min. (GC) | |
| C18H38 | |
| 5 mL | |
| 0.777 | |
| Solubility in water: insoluble. Other solubilities: soluble in ethanol, acetone and diethyl ether | |
| CCCCCCCCCCCCCCCCCC | |
| 254.5 | |
| CHEBI:32926 | |
| 99+% | |
| Octadecane |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
GHS Signal Word: Danger
EINECSNumber : 209-790-3
RUO – Research Use Only