missing translation for 'onlineSavingsMsg'
Learn More

Octadecane, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014793
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014793 1 g
111014794 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014793 Supplier Merck Emd Millipore Supplier No. 746911G
Petroleum

Specifications

Ignition Point 455°F
CAS 593-45-3
Vapor Pressure 1 mmHg (119°C)
Vapor Density 8.8 (vs. air)
Density 0.777 g/mL (at 25°C)
Flash Point 166°C
Physical Form Neat
Linear Formula CH3(CH2)16 CH3
MDL Number MFCD00009007
Melting Point 26°C to 29°C; 28°C to 30°C
Boiling Point 317°C
Type Analytical Standard
Quantity 1 g
Formula Weight 254.49
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less