missing translation for 'onlineSavingsMsg'
Learn More

Octadecafluorodecahydronaphthalene 95.0+%, TCI America™

Catalog No. P083725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P083725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P083725G Supplier TCI America Supplier No. P083725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 306-94-5
Molecular Formula C10F18
Molecular Weight (g/mol) 462.081
MDL Number MFCD00010626
InChI Key UWEYRJFJVCLAGH-UHFFFAOYSA-N
Synonym perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin
PubChem CID 9386
ChEBI CHEBI:38848
IUPAC Name 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene
SMILES C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F

Specifications

Color Colorless
Boiling Point 142°C
Quantity 25 g
Formula Weight 462.08
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Octadecafluorodecahydronaphthalene
EINECSNumber (4)-1176
RTECSNumber QJ3175000
TSCA Yes