missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OCT6 Monoclonal Antibody (KT110), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555998
Description
POU3F1 is a member of the POU domain family of proteins, and regulates events during neurogenesis and myelination.Specifications
| OCT6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P20267, P21952 | |
| POU3F1 | |
| Recombinant mouse Oct6 (proprietary range information). | |
| 200 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| KT110 | |
| Unconjugated | |
| POU3F1 | |
| 6-Oct; LOW QUALITY PROTEIN: POU domain, class 3, transcription factor 1; Oct6; Oct-6; octamer binding factor oct6; octamer-binding protein 6; Octamer-binding transcription factor 6; octoamer transcription factor-6; octomer-6; Otf6; OTF-6; POU class 3 homeobox 1; POU domain transcription factor SCIP; POU domain, class 3, transcription factor 1; POU3F1; Scip; suppressed cAMP-inducible POU; Test1; Testes-1; Tst1; Tst-1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 18991, 192110 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |