missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OCT1 (POU2F1) Recombinant Rabbit Monoclonal Antibody (13H18L58)

Catalog No. 502090064
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502090064 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502090064 Supplier Invitrogen™ Supplier No. 702832

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Bovine, Cat. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

OCT1 (POU2F1) is a ubiquitously expressed transcription factor that binds to the octamer motif (5'-ATTTGCAT-3') and activates the promoters of the genes for some small nuclear RNAs (snRNA) and of genes such as those for histone H2B and immunoglobulins. It modulates transcription transactivation by NR3C1, AR and PGR. In case of human herpes simplex virus (HSV) infection, POU2F1 forms a multiprotein-DNA complex with the viral transactivator protein VP16 and HCFC1 thereby enabling the transcription of the viral immediate early genes.

Specifications

Antigen OCT1 (POU2F1)
Chemical Name or Material Silver Nitrate
Applications ChIP Assay, Western Blot, Immunocytochemistry
Color Colorless to Tan
CAS 7761-88-8, 64-17-5
Classification Recombinant Monoclonal
CAS Min % 2.04, 96.0
Clone 13H18L58
CAS Max % 2.12, 100.0
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene POU2F1
Gene Accession No. P14859
Gene Alias 2810482H01Rik; NF-A1; OCT1; oct-1; oct-1B; octamer binding protein 1; octamer-binding protein 1; Octamer-binding transcription factor 1; Octamer-binding transcription factor-1; Otf1; OTF-1; POU class 2 homeobox 1; POU domain, class 2, transcription factor 1; Pou2f1; ubiquitous transcription factor
Gene Symbols POU2F1
Host Species Rabbit
Immunogen Protein corresponding to human OCT-1 [aa1-aa200].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Molecular Formula AgNO3
Primary or Secondary Primary
MDL Number MFCD00003414
Gene ID (Entrez) 5451
Target Species Human
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Form Liquid
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
Isotype IgG
PubChem CID 24470
ChEBI CHEBI:32130
Grade Laboratory
Packaging Amber Poly Bottle
Solvent or Matrix Denatured Alcohol
Physical Form Liquid
Concentration or Composition Notes 0.100Normal
Show More Show Less