missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ochratoxin A-d5 Solution, 10 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 666236-28-8 |
| Flash Point | 2°C |
| Linear Formula | C20D5H13 ClNO6 |
| MDL Number | MFCD09265279 |
| Synonym | Ochratoxin A-(phenyl-d5) solution |
| Type | Analytical Standard |
| Quantity | 2 mL |
| Formula Weight | 408.84 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 10 μg/mL in acetonitrile |