Learn More
Medicom™ Ocean Pacific™ NeoNatural Powder-Free Textured Chloroprene Medical Examination Gloves
Ocean Pacific™ NeoNatural Powder-Free Textured Chloroprene Medical Examination Gloves. Polychloroprene provides reliable protection, comfort and dexterity. This versatile glove stands out in green. Textured fingertips for a secure grip, even when wet. Chemo tested and chemical resistant.
Supplier: Medicom™ 1228B
Description
- Polychloroprene combines the softness of latex with the strength of nitrile.
- Textured fingertips, chemo tested and chemical resistant.
- Make a bold statement with an exquisite emerald green color.
- Ensure a secure grip for better instrument control under both wet and dry conditions with textured fingertips.
- Thin fit and lightweight to conform perfectly to hands for increased comfort.
Specifications
| No | |
| Healthcare, Industrial, Laboratory | |
| Green | |
| Fingertip Textured | |
| Powder Free | |
| 4 mil | |
| 240 mm |
| No | |
| 1.5 | |
| Single-use | |
| Yes | |
| Small | |
| Yes |