missing translation for 'onlineSavingsMsg'
Learn More

Oakton™ Testr PCTS 50 Waterproof Pocket pH/Cond/TDS/Salinity Tester

Catalog No. 11747329
Change view
Click to view available options
Range (pH):
-1 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range (pH)
11747329 -1 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11747329 Supplier Oakton™ Supplier No. WD3563435

Includes

Protective plastic storage case, lanyard, and batteries.

Great for water, wastewater, laboratory or plant use, these multiparameter testers feature long-life pH electrodes with a wide range of sample compatibility. Ideal for pH, conductivity, TDS, salinity, and temperature testing.

  • Combine measurement capabilities of three testers in one convenient pocket-sized unit
  • Up to 3 point pH calibration with NIST or USA auto buffer recognition
  • Conductivity, TDS, and salinity feature auto ranging based on the concentration of your sample
  • Redesigned leakproof cap functions as a sample/calibration cup, hydrating cup, and stabilizing base to stand the meter upright

Warranty

1 Year

Specifications

Accuracy (Conductivity) ±1% full scale
Accuracy (pH) ±0.01
Accuracy (TDS) ±1% full scale
Backlight Option Yes
Battery Life 150+ hours
Battery Type 4 AAA Alkaline
Calibration Up to 3 points
Calibration Points 3
Channels 1
Display Type Graphics, Dot Matrix 80 x 100 pixel
IP Rating IP67
Measurement Modes pH, Conductivity, TDS, Salinity, Temperature
Range (Conductivity) 0 to 20.00 mS
Range (pH) -1 to 15
Range (Salinity) 0 to 10.00 ppt
Range (TDS) 0 to 10.00 ppt
Resolution (pH) 0.01
Temperature Range (Metric) 0°C to 60°C
For Use With (Application) Water quality testing, buffer and reagent production, electrochemistry
Includes Protective plastic storage case, lanyard, and 4 AAA batteries.
Keypad Membrane
Languages English
Memory Hold Data
Model 50 Series
Probe Type Integral multiparameter probe
Resolution Salinity: 0.10 ppt
Resolution (Conductivity) 0.1 μS, 1 μS; 0.01 mS
Resolution (TDS) 0.1 ppm, 1 ppm, 0.01 ppt
Stability Checkmark indicator
Temperature Accuracy ±0.9°F (0.5°C) from 32 to 122°F (0 to 50°C); ±1.8°F (1.0°C) from 122 to 140°F (50 to 60°C)
Temperature Compensation Automatic
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C
Temperature Range (English) 32°F to 140°F
Temperature Resolution 0.1°F/°C
Warranty 1 Year
Show More Show Less