missing translation for 'onlineSavingsMsg'
Learn More

Oakton™ pH/Ion Benchtop Ion 700 Meter

Catalog No. 13620746
Change view
Click to view available options
Range (pH):
0 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range (pH)
13620746 0 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13620746 Supplier Oakton™ Supplier No. 3541923

Includes

Meter, removable electrode holder, AC adapter

Dual-display benchtop meter for measuring pH, mV, direct ion concentration

  • Features 0.1 mV resolution for ORP measurements
  • Display ppm and mV electrode output in the ion mode simultaneously
  • Selectable buffer sets, five-point pH calibration with auto buffer recognition
  • Manual or automatic temperature compensation from 32° to 212°F (0 to 100°C)
  • With many ion-selective electrodes and built in memory function
  • Built-in electrode stand
  • Stores up to 100 ion, mV, or pH readings
  • Hold function, “Ready” indicator, diagnostic messages
  • Display: 3.25 x 2.87 in. (7.3 x 6.2cm)

Certifications

CE marked, ISO 9001:2000

Warranty

3 year warranty (meter only)

Specifications

Accuracy (mV) ±0.2 within 199.9mV, ±2 beyond 200mV
Accuracy (pH) ±0.01
Calibration Points 5
Certifications/Compliance ISO 9001 2000, CE
Detection Range -2 to +16pH
Range (mV) ±2000 mV
Range (pH) 0 to 14
Resolution (pH) 0.01
For Use With (Application) pH, Ion concentration
Includes Meter, removable electrode holder, AC adapter
Probe Type ATC
Resolution (mV) ±0.1 from 199.9mV, ±1 beyond 199.9mV
Temperature Accuracy ±0.3°C
Temperature Range (English) 41°F to 113°F
Temperature Resolution 0.1°C
Warranty 3 Years
Weight (English) 1.4 lb.
Weight (Metric) 0.6 kg
Voltage 120/240 V
Show More Show Less