missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Calibration Buffer Bottles, Oakton™
Description
- pH buffer solutions allow the user to adjust their system to read accurate measurements
- Store buffers away from direct sunlight.
Order Info
Shipping weight: 1-1/4 lbs. (0.6kg).
Certifications
NIST traceable, Certified to NIST traceable standards.
Specifications
Specifications
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 1.68 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | pH Buffer |
| Grade | Not Available |