missing translation for 'onlineSavingsMsg'
Learn More

Oakton™ PC 700 pH/Conductivity Meter

Catalog No. 13620747
Change view
Click to view available options
Range (pH):
-2 to 16
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range (pH)
13620747 -2 to 16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13620747 Supplier Oakton™ Supplier No. 3541320

Includes

Electrode holder, handy slide-out instruction card, AC adapter

Benchtop meter with advanced setup mode for measuring pH, mV, conductivity, TDS and temperature

  • Easy toggle between pH, mV, conductivity and TDS reading
  • Dual display
  • With pH auto buffer recognition
  • Five-point calibration for pH, conductivity or TDS
  • Memory: up to 100 data sets
  • Selectable auto or manual ranging for conductivity, TDS measurements, automatic end point
  • Manual or automatic temperature compensation from 0° to 100°C (32 to 212°F)
  • Hold function, “Ready” indicator, diagnostic error messages
  • Display: 3.25 x 2.87 in. (8.3 x 6.2cm)
  • Selectable conductivity cell constant (K): 0.1, 1.0, 10.0
  • Adjustable conductivity temperature coefficient: 0.0 to 10.0% per °C
  • Adjustable conductivity-to-TDS conversion factor:0.4 to 1.0
  • Adjustable Normalization temperature: 15 to 30°C
For water quality testing

Certifications

CE marked, ISO 9001:2000

Warranty

3 year warranty (meter only)

Specifications

Accuracy (mV) ±2000 mV
Accuracy (pH) ±0.01
Calibration Points (Conductivity) 1 Point/Range (up to 5)
Calibration Points (pH) Up to 5 points (USA: 1.68, 4.01, 7, 10, 12.45, NIST: 1.68, 4.01, 6.86, 9.18, 12.45)
Certifications/Compliance CE, ISO 9001 2000
Data Points 100
Detection Range -2 to +16pH
Range (Conductivity) 0 to 19.99 μS, 0 to 199.9 μS, 0 to 1999 μS, 0 to 19.99 mS, 0 to 200 mS
Range (mV) ±2000 mV
Range (pH) -2 to 16
Range (TDS) 0 to 9.99 ppm, 10 to 99.9 ppm, 100 to 999 ppm, 1 to 9.99 ppt, 10 to 99.9 ppt, 100 to 200 ppt
Resolution (pH) 0.01
Temperature Range (Metric) 0°C to 100°C
Includes Electrode holder, handy slide-out instruction card, AC adapter
Resolution (Conductivity) 0.01μS, 0.1μS, 1μS, 0.01mS, 0.1mS
Resolution (mV) ±0.1 from 199.9mV, ±1 beyond 200mV
Resolution (TDS) 0.01ppm, 0.1ppm, 1ppm, 0.01ppt, 0.1ppt
Temperature Accuracy ±0.5°C
Temperature Resolution 0.1°C
Warranty 3 Years
Weight (English) 1.25 lb.
Weight (Metric) 0.6 kg
Voltage 100/240 V
Show More Show Less