missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oakton™ ORPTestr 50 Waterproof Pocket ORP Tester
Oakton ORP 50 Waterproof TestrTM features a replaceable double-junction platinum electrode. Ideal for checking dirty water, heavy metals, sulfides, and organics. ValoxTMelectrode body resists most chemicals.
Supplier: Oakton™ 3563445
Includes: Protective plastic storage case, lanyard, and 4 AAA batteries.
Description
- Redesigned leakproof cap functions as a sample cup, hydrating cup, and as a stabilizing base to stand meter upright
- Replaceable, double-junction electrode sensors save time and money
- Easy-to-navigate interface for quick setup and calibration
Warranty
1 Year
Specifications
| ±2 mV | |
| 250+ hr. | |
| 1 point | |
| 1 | |
| IP67 | |
| ±1000 mV | |
| Water quality testing, buffer and reagent production, electrochemistry | |
| Membrane | |
| Hold Data | |
| Integral electrode | |
| ±1000 mV | |
| Checkmark indicator | |
| 41°F to 113°F (ambient) | |
| 32°F to 140°F | |
| 1 Year |
| Yes | |
| 4 AAA Alkaline | |
| 1 | |
| Graphics, Dot Matrix 80 x 100 pixel | |
| ORP, Temperature | |
| 0°C to 60°C | |
| Protective plastic storage case, lanyard, and 4 AAA batteries. | |
| English | |
| 50 Series | |
| ORP Meter | |
| 1 mV | |
| Manual | |
| 5°C to 45°C | |
| 0.1°F/°C |