missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAC1 98.0+%, TCI America™
$249.78 - $749.35
Chemical Identifiers
| CAS | 300586-90-7 |
|---|---|
| Molecular Formula | C14H11N3O |
| Molecular Weight (g/mol) | 237.26 |
| MDL Number | MFCD00572806 |
| InChI Key | HWJRIFZDXJKJJN-UHFFFAOYSA-N |
| Synonym | oac-1, n-1h-pyrrolo 2,3-c pyridin-5-ylbenzamide, n-1h-pyrrolo 2,3-c pyridin-5-yl benzamide, cambridge id 5236727, oct4-activating compound 1, benzamide,n-1h-pyrrolo 2,3-c pyridin-5-yl, benzamide, n-1h-pyrrolo 2,3-c pyridin-5-yl, n-1h-pyrrolo 2,3-c pyridin-5-yl-benzamide |
| PubChem CID | 789882 |
| IUPAC Name | N-{1H-pyrrolo[2,3-c]pyridin-5-yl}benzamide |
| SMILES | O=C(NC1=NC=C2NC=CC2=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 300586-90-7 | |
| 237.26 | |
| HWJRIFZDXJKJJN-UHFFFAOYSA-N | |
| 789882 | |
| O=C(NC1=NC=C2NC=CC2=C1)C1=CC=CC=C1 |
| C14H11N3O | |
| MFCD00572806 | |
| oac-1, n-1h-pyrrolo 2,3-c pyridin-5-ylbenzamide, n-1h-pyrrolo 2,3-c pyridin-5-yl benzamide, cambridge id 5236727, oct4-activating compound 1, benzamide,n-1h-pyrrolo 2,3-c pyridin-5-yl, benzamide, n-1h-pyrrolo 2,3-c pyridin-5-yl, n-1h-pyrrolo 2,3-c pyridin-5-yl-benzamide | |
| N-{1H-pyrrolo[2,3-c]pyridin-5-yl}benzamide |
Specifications
| 300586-90-7 | |
| C14H11N3O | |
| 10 mg | |
| HWJRIFZDXJKJJN-UHFFFAOYSA-N | |
| N-{1H-pyrrolo[2,3-c]pyridin-5-yl}benzamide | |
| 789882 | |
| ≥98.0% (GC) | |
| OAC1 |
| White-Yellow | |
| MFCD00572806 | |
| oac-1, n-1h-pyrrolo 2,3-c pyridin-5-ylbenzamide, n-1h-pyrrolo 2,3-c pyridin-5-yl benzamide, cambridge id 5236727, oct4-activating compound 1, benzamide,n-1h-pyrrolo 2,3-c pyridin-5-yl, benzamide, n-1h-pyrrolo 2,3-c pyridin-5-yl, n-1h-pyrrolo 2,3-c pyridin-5-yl-benzamide | |
| O=C(NC1=NC=C2NC=CC2=C1)C1=CC=CC=C1 | |
| 237.26 | |
| 237.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer