accessibility menu, dialog, popup

UserName

OHNNHN

CAS RN 300586-90-7

IUPAC Name: N-{1H-pyrrolo[2,3-c]pyridin-5-yl}benzamide
Synonyms: N-{1H-pyrrolo[2,3-c]pyridin-5-yl}benzamide
Molecular Weight (g/mol): 237.26
Molecular Formula: C14H11N3O
InChi Key: HWJRIFZDXJKJJN-UHFFFAOYSA-N
SMILES: O=C(NC1=NC=C2NC=CC2=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results